Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Cresol Red, Sodium Salt
SCI_ED

Catalog No. S25272 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25272 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S25272

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 62625-29-0
Molecular Formula C21H17O5S
Synonym O-Cresolsulfonephthalein

Specifications

Linear Formula C21H17NaO5S
Formula Weight 404.4
Packaging Powder
Quantity 1 g
Physical Form Solid
Chemical Name or Material Cresol Red, Sodium Salt
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.