Learn More
Abcam Cresol red sodium salt, pH indicator dye, 25G
Catalog No. 502704725
Encompass_Preferred
MW 404.4 g/mol. pH indicator with color changes for acidic solutions of pH 0.2 (red) to pH 1.8 (yellow), and for basic solutions of pH 7.2 (yellow) to pH 8.8 (red). Used in many common molecular biology reactions in place of other loading dyes. It does not inhibit Taq polymerase to the same degree as other common loading dyes.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4725 Supplier Abcam Supplier No. AB14632025G
MW 404.4 g/mol. pH indicator with color changes for acidic solutions of pH 0.2 (red) to pH 1.8 (yellow), and for basic solutions of pH 7.2 (yellow) to pH 8.8 (red). Used in many common molecular biology reactions in place of other loading dyes. It does not inhibit Taq polymerase to the same degree as other common loading dyes.
The product is subject to the following: Abcam Restricted Use Statement