Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cresol Red-Thymol Blue TS, Ricca Chemical
SDP

Catalog No. 24554 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
2455-4 120 mL Natural Poly Bottle
2455-16 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 2455-4 Supplier Ricca Chemical Company Supplier No. 24554
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Cresol Red-Thymol Blue
Name Note Test Solution
CAS 64-17-5, 7732-18-5, 67-56-1, 76-61-9, 62625-29-0
CAS Min % 69.0, 28.79, 0.09, 0.01
CAS Max % 72.0, 29.97, 0.09, 0.01
Grade Indicator
Packaging Natural Poly Bottle
Color Red
Quantity 120 mL
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.