Learn More
Abcam Cresol red, Triarylmethane dye, 25G
Catalog No. 502704727
Encompass_Preferred
MW 382.43 g/mol. Triarylmethane dye commonly used with lead acetate for the detection of urease after polyacrylamide gel electrophoresis (PAGE). Also used as a pH indicator and as a tracking dye in DNA, RNA (agarose) and protein (polyacrylamide) electrophoresis. In agarose, Cresol Red runs with an apparent molecular size of approximately 125 bp DNA. Cresol Red does not inhibit Taq polymerase.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4727 Supplier Abcam Supplier No. AB14631925G
MW 382.43 g/mol. Triarylmethane dye commonly used with lead acetate for the detection of urease after polyacrylamide gel electrophoresis (PAGE). Also used as a pH indicator and as a tracking dye in DNA, RNA (agarose) and protein (polyacrylamide) electrophoresis. In agarose, Cresol Red runs with an apparent molecular size of approximately 125 bp DNA. Cresol Red does not inhibit Taq polymerase.
The product is subject to the following: Abcam Restricted Use Statement