Learn More
MARSHALL SCIENTIFIC Crest P1100HT-45 Digital Ultrasonic Cleaner
SDP
Catalog No. NC2591241
Encompass_Preferred
The Crest P1100HT-45 Digital Ultrasonic Cleaner has a 3.25 gallon tank size and utilizes a digital heater to set temperature and total run time.
Catalog No. NC2591241 Supplier MARSHALL SCIENTIFIC Supplier No. 1100PH0451
The Crest P1100HT-45 Digital Ultrasonic Cleaner has a 3.25 gallon tank size and utilizes a digital heater to set temperature and total run time.