Learn More
MARSHALL SCIENTIFIC Crest P230HT-45 Digital Ultrasonic Cleaner
SDP
Catalog No. NC2590790
Encompass_Preferred
The Crest P230HT-45 Digital Ultrasonic Cleaner has a 0.75 gallon tank size and utilizes a digital heater to set temperature and total run time.
Catalog No. NC2590790 Supplier MARSHALL SCIENTIFIC Supplier No. 0230PH0451
The Crest P230HT-45 Digital Ultrasonic Cleaner has a 0.75 gallon tank size and utilizes a digital heater to set temperature and total run time.