Learn More
MARSHALL SCIENTIFIC Crest P2600HT-45 Ultrasonic cleaner
SDP
Catalog No. NC2591247
Encompass_Preferred
The Crest P2600HT-45 Digital Ultrasonic Cleaner has a 7 gallon tank size and utilizes a digital heater to set temperature and total run time.
Catalog No. NC2591247 Supplier MARSHALL SCIENTIFIC Supplier No. 2600PH0451
The Crest P2600HT-45 Digital Ultrasonic Cleaner has a 7 gallon tank size and utilizes a digital heater to set temperature and total run time.