Learn More
MARSHALL SCIENTIFIC Crest P360HTPC-45 Digital Ultrasonic Cleaner w/ Power Control
SDP
Catalog No. NC2590799
Encompass_Preferred
The Crest P360HTPC-45 Digital Ultrasonic Cleaner w/ Power Control has a 1 gallon tank size and utilizes a digital heater to set temperature and total run time.
Catalog No. NC2590799 Supplier MARSHALL SCIENTIFIC Supplier No. 0360PD0451
The Crest P360HTPC-45 Digital Ultrasonic Cleaner w/ Power Control has a 1 gallon tank size and utilizes a digital heater to set temperature and total run time.