Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cresyl Blue, MilliporeSigma™

Catalog No. MR0302170 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MR0302170 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MR0302170 Supplier MilliporeSigma Supplier No. R0302170

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 81029-05-2
Synonym Gentian Violet

Specifications

Formula Weight 321.32
Grade Certified Biological Stain
Quantity 100 mL
Physical Form Crystalline
Chemical Name or Material Cresyl Blue, HARLECO™
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.