Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cresyl Blue, HARLECO™, MilliporeSigma™

Catalog No. M16312 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16312 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M16312 Supplier MilliporeSigma Supplier No. 16312

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 81029-05-2
Molecular Formula C17H20N3OCl·0.5ZnCl2
Synonym Brilliant Blue C

Specifications

Formula Weight 385.96
Color Green
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Cresyl Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.