Learn More
Medchemexpress LLC Cresyl diphenyl phosphate | 26444-49-5 | MFCD00014920 | 97.0% | 340.31 g·mol⁻¹ | C19H17O4P | 100 G
SDP
Catalog No. 5000280906
Encompass_Preferred
Cresyl diphenyl phosphate is an organophosphate research chemical (CAS 26444-49-5) used in biochemical and developmental biology studies; it disrupts lipid homeostasis in zebrafish embryos. The material is supplied as a colorless to light yellow liquid with a reported purity of 97.0% and a molecular weight of 340.31 g·mol⁻¹.
  • Research-grade organophosphate for laboratory studies.
  • Disrupts lipid homeostasis in zebrafish embryos, useful for developmental assays.
  • High analytical purity suitable for biochemical and assay applications.
  • Supplied as a liquid with defined storage recommendations to maintain stability.
  • Provided with documented safety and handling guidance for research use only.
Catalog No. 50-002-80906 Supplier Medchemexpress LLC Supplier No. HYW250743100G
Cresyl diphenyl phosphate is an organophosphate research chemical (CAS 26444-49-5) used in biochemical and developmental biology studies; it disrupts lipid homeostasis in zebrafish embryos. The material is supplied as a colorless to light yellow liquid with a reported purity of 97.0% and a molecular weight of 340.31 g·mol⁻¹.
  • Research-grade organophosphate for laboratory studies.
  • Disrupts lipid homeostasis in zebrafish embryos, useful for developmental assays.
  • High analytical purity suitable for biochemical and assay applications.
  • Supplied as a liquid with defined storage recommendations to maintain stability.
  • Provided with documented safety and handling guidance for research use only.