Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cresyl diphenyl phosphate | 26444-49-5 | MFCD00014920 | 97.0% | 340.31 g/mol | C19H17O4P | 25 G
SDP

Catalog No. 5000281634
Encompass_Preferred

Cresyl diphenyl phosphate is an organophosphate compound used as a biochemical assay reagent and in toxicology research; it has been reported to disrupt lipid homeostasis in model organisms.

  • Chemical formula: C19H17O4P.
  • Molecular weight: 340.31 g/mol.
  • Cas number: 26444-49-5.
  • Purity: 97.0%.
  • Recommended storage: pure form -20°C (3 years), 4°C (2 years).
  • Common applications include biochemical assays and studies of lipid metabolism.

Catalog No. 50-002-81634 Supplier Medchemexpress LLC Supplier No. HYW25074325G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cresyl diphenyl phosphate is an organophosphate compound used as a biochemical assay reagent and in toxicology research; it has been reported to disrupt lipid homeostasis in model organisms.

  • Chemical formula: C19H17O4P.
  • Molecular weight: 340.31 g/mol.
  • Cas number: 26444-49-5.
  • Purity: 97.0%.
  • Recommended storage: pure form -20°C (3 years), 4°C (2 years).
  • Common applications include biochemical assays and studies of lipid metabolism.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.