Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cresyl diphenyl phosphate | 26444-49-5 | MFCD00014920 | 97.0% | 340.31 g/mol | C19H17O4P | 50 G
SDP

Supplier:  Medchemexpress LLC HYW25074350G

Encompass_Preferred

Cresyl diphenyl phosphate is an organophosphate research reagent used in biochemical assays and studies of lipid metabolism. It is supplied as a high-purity solid suitable for laboratory use, with reported molecular formula C19H17O4P and molecular weight 340.31 g/mol. The compound is commonly employed in in vitro and in vivo studies examining lipid homeostasis and related pathways.

  • High purity: 97.0%.
  • Molecular formula: C19H17O4P.
  • Molecular weight: 340.31 g/mol.
  • Used in biochemical assays and lipid metabolism research.
  • Available in laboratory-scale pack sizes for research use.

Catalog No. 50-002-80908


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.