Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Cresyl Fast Violet, Certified, C.I. DcW-5 5 GR
SDP

Catalog No. 50980338
Encompass_Preferred

(Cresyl Violet Acetate)
CAS #10510-54-0 C₁₈H₁₅N₃O₃ F.W. 321.34
Solubility: 0.13% water

Vogt's method for nerve cells. A neurological tissue stain. Nissl substance & PAS-positive material. Powers & Clark method for spinal cord and brain with formalin or Bouins fixed.

Catalog No. 50-980-338 Supplier Electron Microscopy Sciences Supplier No. 12780
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Cresyl Violet Acetate)
CAS #10510-54-0 C₁₈H₁₅N₃O₃ F.W. 321.34
Solubility: 0.13% water

Vogt's method for nerve cells. A neurological tissue stain. Nissl substance & PAS-positive material. Powers & Clark method for spinal cord and brain with formalin or Bouins fixed.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.