Learn More
Electron Microscopy Sciences Cresyl Violet Acetate Solution, 0.1% aqueous, 500ml
SDP
Catalog No. 5031945
Encompass_Preferred
Cresyl-violet acetate is used in Nissl staining of the brain and spinal cord. It is also stain for mast cell and cartilage granules.
Catalog No. 50-319-45 Supplier Electron Microscopy Sciences Supplier No. 2608901
Cresyl-violet acetate is used in Nissl staining of the brain and spinal cord. It is also stain for mast cell and cartilage granules.