Learn More
Electron Microscopy Sciences Cresyl Violet Acetate 2% Aqueous 250 ML
SDP
Catalog No. 5031944
Encompass_Preferred
Cresyl Violet Acetate solution is a prepared, ready-to-use, high quality staining solution that saves time and effort and produces consistent uniform results.
Catalog No. 50-319-44 Supplier Electron Microscopy Sciences Supplier No. 2608920
Cresyl Violet Acetate solution is a prepared, ready-to-use, high quality staining solution that saves time and effort and produces consistent uniform results.