Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cresyl violet acetate, 69%, MP Biomedicals™

Catalog No. ICN15072701 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15072701 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15072701 Supplier MP Biomedicals, Inc Supplier No. 0215072701
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Supplied as a very dark, near-black powder with a dye content of 98% for laboratory applications

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10510-54-0
Molecular Formula C18H15N3O3
Molecular Weight (g/mol) 321.34
MDL Number MFCD00013151,MFCD00013151
InChI Key XKOCOMKJPWEOHX-UHFFFAOYSA-M
Synonym Cresyl Fast Violet, 9-amino-5-imino-5H-benzo[a]phenoxazine acetate salt
PubChem CID 44134641
IUPAC Name 8,12-diamino-10λ⁴-oxa-5-azatetraphen-10-ylium acetate
SMILES CC([O-])=O.NC1=CC2=[O+]C3=C(N=C2C=C1)C1=CC=CC=C1C(N)=C3

Specifications

Assay Percent Range 69%
Color Green
Quantity 1 g
Physical Form Powder
Formula Weight 321.3
Grade Certified Biological Stain
Chemical Name or Material Cresyl violet acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.