Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cresyl violet (acetate), For Microscopy, Certistain™, MilliporeSigma™

Catalog No. M1052350025 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1052350025 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1052350025 Supplier MilliporeSigma Supplier No. 1.05235.0025

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10510-54-0
Molecular Formula C18H15N3O3
Molecular Weight (g/mol) 321.34
MDL Number MFCD00013151,MFCD00013151
InChI Key XKOCOMKJPWEOHX-UHFFFAOYSA-M
PubChem CID 44134641
IUPAC Name 8,12-diamino-10λ⁴-oxa-5-azatetraphen-10-ylium acetate
SMILES CC([O-])=O.NC1=CC2=[O+]C3=C(N=C2C=C1)C1=CC=CC=C1C(N)=C3

Specifications

Formula Weight 321.33 (molar mass)
Odor Weak odor
Packaging Glass bottle
Color Dark Green
Quantity 25 g
Physical Form Solid
Chemical Name or Material Cresyl violet (acetate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.