Learn More
Electron Microscopy Sciences Cresyl Violet Acetate Solution 0.1% 250 ML
SDP
Catalog No. 5031887
Encompass_Preferred
Used in Kluver-Barrera Method for Myelin and Nerve Calls for relation of nerve cells to neutroglia, etc,. References: Kluver, H., and Barrera, E.: A method for the combined staining of cells and fibers in the nervous system. J. Nueropath. Exp. Neurol. 12:400 – 403, 1953 and AFIP, Lab Methods in Histotechnology, ed. Edna B. Prophet, Bob Mills. Jacquelyn B. Arrmyl, Leslie H. Sobin, M.D. Wash, D.C. American Registry of Pathology, c. 1992, p. 94-95.
Catalog No. 50-318-87 Supplier Electron Microscopy Sciences Supplier No. 2668102
Used in Kluver-Barrera Method for Myelin and Nerve Calls for relation of nerve cells to neutroglia, etc,. References: Kluver, H., and Barrera, E.: A method for the combined staining of cells and fibers in the nervous system. J. Nueropath. Exp. Neurol. 12:400 – 403, 1953 and AFIP, Lab Methods in Histotechnology, ed. Edna B. Prophet, Bob Mills. Jacquelyn B. Arrmyl, Leslie H. Sobin, M.D. Wash, D.C. American Registry of Pathology, c. 1992, p. 94-95.