Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cresyl Violet Acetate, Spectrum™ Chemical
SDP

Catalog No. 18620493 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-493 1 g
18-620-494 5 g
18-620-495 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-620-493 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. CR1251GM
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10510-54-0
Molecular Weight (g/mol) 321.34

Specifications

Quantity 1 g
Packaging Amber Glass
Grade Ungraded
Chemical Name or Material Cresyl Violet Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.