Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cresyl Violet Acetate, Spectrum™ Chemical
SDP

Catalog No. 18620494 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-494 5 g
18-620-493 1 g
18-620-495 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-620-494 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. CR1255GM
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10510-54-0
Molecular Weight (g/mol) 321.34

Specifications

Quantity 5 g
Packaging Amber Glass
Molecular Formula C18H15N3O3
InChI Key XKOCOMKJPWEOHX-UHFFFAOYSA-M
SMILES CC([O-])=O.NC1=CC2=[O+]C3=C(N=C2C=C1)C1=CC=CC=C1C(N)=C3
IUPAC Name 8,12-diamino-10λ⁴-oxa-5-azatetraphen-10-ylium acetate
Grade Ungraded
Chemical Name or Material Cresyl Violet Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.