Learn More
Electron Microscopy Sciences Cresyl Violet Acetate Working Solution 250 ML
SDP
Catalog No. 5031882
Encompass_Preferred
Used in Vogt's Method for Nerve Cell Products. AFIP, Manual of Histological Staining Methods, 3rd Ed., Ed. L. Luna: New York: McGraw-Hill Publications, c. 1968, p. 212
Catalog No. 50-318-82 Supplier Electron Microscopy Sciences Supplier No. 266711A
Used in Vogt's Method for Nerve Cell Products. AFIP, Manual of Histological Staining Methods, 3rd Ed., Ed. L. Luna: New York: McGraw-Hill Publications, c. 1968, p. 212