Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Cresyl Violet perchlorate, 98%, pure

Catalog No. AC405775000
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC405775000 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC405775000 Supplier Thermo Scientific Chemicals Supplier No. 405775000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 41830-80-2 | C16H12ClN3O5 | 361.738 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41830-80-2
Molecular Formula C16H12ClN3O5
Molecular Weight (g/mol) 361.738
MDL Number MFCD00012671
InChI Key HLHUTAPXZITNPJ-UHFFFAOYSA-N
Synonym Oxazine 9 perchlorate
PubChem CID 621529
IUPAC Name 5-iminobenzo[a]phenoxazin-9-amine;perchloric acid
SMILES C1=CC=C2C(=C1)C(=N)C=C3C2=NC4=C(O3)C=C(C=C4)N.OCl(=O)(=O)=O

Specifications

Melting Point 331.0°C to 333.0°C
Fluorescence Max 615nm (in methanol)
Infrared Spectrum Authentic
Lambda Max 593nm (in methanol)
Molar Absorption Coefficient 68000L/(mole.cm) (lambda max.)
Packaging Glass bottle
Suitability (for use as a laser dye) passes
Assay Percent Range 98%
UV_Vis 98.5% min. (on dry substance)
Color Green
Solubility Information Solubility in water: insoluble.
Quantity 500 mg
Physical Form Powder
Formula Weight 361.73
Percent Purity 98%
Grade Pure
Chemical Name or Material Cresyl violet perchlorate, 98%
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.