Learn More
Fd Neurotechnologies Inc FD Cresyl Violet Solution™ (Double Strength)
SDP
Catalog No. NC0527138
Encompass
FD Cresyl Violet Solution™, made from cresyl violet acetate certified by the BIOLOGICAL STAIN COMMISSION and with a step-by-step user manual printed on the bottle, is specifically designed for easy use in all types of laboratories. This solution is formulated and extensively tested for the staining of nerve tissue. Cresyl violet imparts a violet color to nuclear chromatin and Nissl substances. This solution is excellent for visualizing neuronal cell bodies and glial nuclei.
Catalog No. NC0527138 Supplier Fd Neurotechnologies Inc Supplier No. PS1022
FD Cresyl Violet Solution™, made from cresyl violet acetate certified by the BIOLOGICAL STAIN COMMISSION and with a step-by-step user manual printed on the bottle, is specifically designed for easy use in all types of laboratories. This solution is formulated and extensively tested for the staining of nerve tissue. Cresyl violet imparts a violet color to nuclear chromatin and Nissl substances. This solution is excellent for visualizing neuronal cell bodies and glial nuclei.