Learn More
Fd Neurotechnologies Inc CRESYL VIOLET SOLUTION 500ML
SDP
Catalog No. NC0527137
Encompass
FD Cresyl Violet Solution™, made from cresyl violet acetate certified by the BIOLOGICAL STAIN COMMISSION and with a step by step user manual printed on the bottle, is specifically designed for easy use in all types of laboratories. This solution is formulated and extensively tested for the staining of nerve tissue. Cresyl violet imparts a violet color to nuclear chromatin and Nissl substances. This solution is excellent for visualizing neuronal cell bodies and glial nuclei. FD Cresyl Violet Solution™ is formulated for the staining of both neurons and glial cells. This solution can be used with frozen or paraffin-embedded tissue sections fixed with any fixative (formalin preferred). 500ml
Catalog No. NC0527137 Supplier Fd Neurotechnologies Inc Supplier No. PS1021
FD Cresyl Violet Solution™, made from cresyl violet acetate certified by the BIOLOGICAL STAIN COMMISSION and with a step by step user manual printed on the bottle, is specifically designed for easy use in all types of laboratories. This solution is formulated and extensively tested for the staining of nerve tissue. Cresyl violet imparts a violet color to nuclear chromatin and Nissl substances. This solution is excellent for visualizing neuronal cell bodies and glial nuclei. FD Cresyl Violet Solution™ is formulated for the staining of both neurons and glial cells. This solution can be used with frozen or paraffin-embedded tissue sections fixed with any fixative (formalin preferred). 500ml