Learn More
Abcam Cresyl Violet Stain Solution (0.1%), 500ML
Catalog No. NC1912951
Encompass_Preferred
Cresyl violet stain solution (0.1%) is a staining solution for visualizing Nissl substance in neurons on formalin-fixed, paraffin-embedded tissue. - Stains Nissl granules violet and neuroglia nuclei slightly bluer - Ideal for cerebral cortex tissue analysis
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1912951 Supplier Abcam Supplier No. AB246816500ML
Cresyl violet stain solution (0.1%) is a staining solution for visualizing Nissl substance in neurons on formalin-fixed, paraffin-embedded tissue. - Stains Nissl granules violet and neuroglia nuclei slightly bluer - Ideal for cerebral cortex tissue analysis
The product is subject to the following: Abcam Restricted Use Statement