Learn More
MCR Safety Crews™ Tomahawk™ Safety Glasses
Catalog No. 19034308 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Frame Color:
Black
Blue
Red, White, Blue
Lens Tint:
Clear
Gray
Lens Coating:
Scratch-resistant
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint Lens Coating
19-034-308 Red, White, Blue Clear -
18-999-1905 Black Clear Scratch-resistant
19-034-304 Black Clear -
18-999-1909 Red, White, Blue Clear Scratch-resistant
18-999-1906 Black Gray -
18-999-1907 Blue Clear Scratch-resistant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-034-308 Supplier MCR Safety Supplier No. TK130AF
This item is not returnable. View return policy
The Tomahawk™ is an affordable alternative for providing comfort, protection and style. Features include a stylish wrap-around design and one-piece polycarbonate lens with integral sideshields.
  • High brow guard for extra protection and unimpeded view
  • Single lens design with integral side shields for greater orbital seal
  • Spatula temple with extendable adjustment
  • Rugged polycarbonate lens filters 99.9% of harmful U.V. rays
Compliance
Spectacles meet ANSI Z87.1-1989 standards.
Specifications
Certifications/Compliance Meet ANSI Z87.1-1989 safety standards
Lens Tint Clear
Frame Color Red, White, Blue
Lens Material Polycarbonate
Frame Material Nylon