Learn More
Medchemexpress LLC HY-P1533A 1mg Medchemexpress, CRF, bovine (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1533A1MG
Encompass_Preferred
Medchemexpress, HY-P1533A 1mg CRF, bovine (TFA) CAS: CRF, bovine (TFA) is a potent agonist of CRF receptor , and displaces [ 125 I-Tyr]ovine CRF with a K i of 3.52 nM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4222