Learn More
Echelon Biosciences Research Labs CRF, human, rat; Corticotropin Releasing Factor - 1 mg, CAS#: 86784-80-7
SDP
Supplier:  Echelon Biosciences Research Labs 351251MG
Encompass_Preferred
CRF, human, rat; Corticotropin Releasing Factor - 1mg , Ser-Glu-Glu-Pro-Pro-Ile-Ser-Leu-Asp-Leu-Thr-Phe-His-Leu-Leu-Arg-Glu-Val-Leu-Glu-Met-Ala-Arg-Ala-Glu-Gln-Leu-Ala-Gln-Gln-Ala-His-Ser-Asn-Arg-Lys-Leu-Met-Glu-Ile-Ile-NH2 Category/Application: Neuropeptides Hormones This item is non returnable
Catalog No. 50-190-8417