Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC cRGDfK-thioacetyl ester TFA | 99.3% | 719.81 | 1 MG
SDP

Catalog No. 5000396955
Encompass_Preferred

cRGDfK-thioacetyl ester TFA is a bioactive polypeptide molecule that exhibits selective affinity for integrins. It can be utilized to modify NIR fluorescent probes for cancer targeting imaging.

  • Bioactive polypeptide molecule
  • Exhibits selective affinity for integrins
  • Modifies NIR fluorescent probes for cancer targeting imaging
  • Appearance: Solid
  • Color: White to off-white
  • Soluble in DMSO at concentrations ≥ 100 mg/mL

Catalog No. 50-003-96955 Supplier Medchemexpress LLC Supplier No. HYP5287A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

cRGDfK-thioacetyl ester TFA is a bioactive polypeptide molecule that exhibits selective affinity for integrins. It can be utilized to modify NIR fluorescent probes for cancer targeting imaging.

  • Bioactive polypeptide molecule
  • Exhibits selective affinity for integrins
  • Modifies NIR fluorescent probes for cancer targeting imaging
  • Appearance: Solid
  • Color: White to off-white
  • Soluble in DMSO at concentrations ≥ 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.