Learn More
Aviva Systems Biology Corp CRHR1 Antibody - N-terminal region (ARP63606_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63606_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CRHR1. It was validated on Western Blot. . Alternate names: CRF-R, CRF1, CRFR1, CRH-R1h, CRHR, CRHR1L, CRHR1f, CRF-R1, CRFR-1, CRH-R1, CRF-R-1, CRH-R-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-027