Learn More
Aviva Systems Biology Corp CRHR2 Antibody - N-terminal region (ARP64081_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64081_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CRHR2. It was validated on Western Blot. . Alternate names: CRFR2, CRF2, CRF-RB, HM-CRFApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-288-168