Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cridanimod | 38609-97-1 | 99.8% | 253.26 | 250 MG
SDP

Catalog No. 5000454043
Encompass_Preferred

Cridanimod is a potent progesterone receptor (PR) activator that works by inducing IFNα and IFNβ expression. It functions as a small-molecule immunomodulator and interferon inducer.

  • Potent progesterone receptor (PR) activator.
  • Induces IFNα and IFNβ expression.
  • Small-molecule immunomodulator.
  • Interferon inducer.
  • Purity: 99.8%.
  • Molecular weight: 253.26.
  • Appearance: Solid, light yellow to yellow.
  • Soluble in DMSO (125 mg/mL).
  • Increases survival time and elevates IFNα and -β levels in athymic mice.
  • Stable as powder for up to 3 years at -20°C, 2 years at 4°C.
  • Stable in solvent for up to 2 years at -80°C, 1 year at -20°C.

Catalog No. 50-004-54043 Supplier Medchemexpress LLC Supplier No. 250MGHYW011890
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cridanimod is a potent progesterone receptor (PR) activator that works by inducing IFNα and IFNβ expression. It functions as a small-molecule immunomodulator and interferon inducer.

  • Potent progesterone receptor (PR) activator.
  • Induces IFNα and IFNβ expression.
  • Small-molecule immunomodulator.
  • Interferon inducer.
  • Purity: 99.8%.
  • Molecular weight: 253.26.
  • Appearance: Solid, light yellow to yellow.
  • Soluble in DMSO (125 mg/mL).
  • Increases survival time and elevates IFNα and -β levels in athymic mice.
  • Stable as powder for up to 3 years at -20°C, 2 years at 4°C.
  • Stable in solvent for up to 2 years at -80°C, 1 year at -20°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.