Learn More
Cayman Chemical CrIdnImododIum salt 10mg
SDP
Catalog No. 502941305
Encompass_Preferred
An inducer of type I interferon production; induces IRF3 phosphorylation, IFN-β production, and NF-κB activation in wild-type, but not in STING mutant, murine macrophages; increases plasma levels of IFN in weanling and adult mice from 112-1,792 mg/kg; protects against viral infection in mouse models of Semliki forest, coxsackie B1, Columbia SK, herpes, and pseudorabies viruses (PD50s = 17-320 mg/kg); increases uterine expression of estrogen and progesterone receptors in ovariectomized rats; reverses tamoxifen-induced decreases in progesterone receptor expression in young rats
Catalog No. 50-294-1305 Supplier Cayman Chemical Supplier No. 2788410 MG
An inducer of type I interferon production; induces IRF3 phosphorylation, IFN-β production, and NF-κB activation in wild-type, but not in STING mutant, murine macrophages; increases plasma levels of IFN in weanling and adult mice from 112-1,792 mg/kg; protects against viral infection in mouse models of Semliki forest, coxsackie B1, Columbia SK, herpes, and pseudorabies viruses (PD50s = 17-320 mg/kg); increases uterine expression of estrogen and progesterone receptors in ovariectomized rats; reverses tamoxifen-induced decreases in progesterone receptor expression in young rats