Learn More
World Precision Instrument Crile Forceps, 14cm, Curved, 12-pack, Disposable
SDP
Catalog No. 502539457
Encompass
Crile hemostatic forceps are atraumatic and non-toothed clamps used to clamp blood vessels, grab tissue or vessels. They are often known as a "snap."
Catalog No. 50-253-9457 Supplier World Precision Instrument Supplier No. 50370112
Crile hemostatic forceps are atraumatic and non-toothed clamps used to clamp blood vessels, grab tissue or vessels. They are often known as a "snap."