Learn More
World Precision Instrument Crile Forceps, 14cm, Straight, 12-pack
SDP
Catalog No. 502539458
Encompass
Crile hemostatic forceps are atraumatic and non-toothed clamps used to clamp blood vessels, grab tissue or vessels. They are often known as a "snap."
Catalog No. 50-253-9458 Supplier World Precision Instrument Supplier No. 50370012
Crile hemostatic forceps are atraumatic and non-toothed clamps used to clamp blood vessels, grab tissue or vessels. They are often known as a "snap."