Learn More
Stoelting™ Crile Forceps
SDP
Catalog No. 10001343 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Curved
Curved, Extra delicate
Straight
Straight, Extra delicate
Size:
5.5 in.
6.25 in.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Size
10-001-343 Straight 6.25 in.
10-001-341 Straight 5.5 in.
10-001-338 Straight, Extra delicate 5.5 in.
10-001-342 Curved 5.5 in.
10-001-340 Curved, Extra delicate 5.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 10-001-343 Supplier Stoelting™ Supplier No. 5211025
Add to Cart
Add to Cart
Hemostatic forceps are used in many surgical procedures to control bleeding or the flow of liquids in tubing.
  • Scissor-like ring handles
  • Ratcheted handles can be locked in multiple positions in order to maintain variable levels of constant pressure
Specifications
For Use With (Application) Bleeding Control
Size 6.25 in.
Model Straight