Learn More
World Precision Instrument Crile Hemostatic Forceps 12cm 1X2 teeth, Titanium
SDP
Catalog No. 502538285
Encompass
Crile hemostatic forceps are atraumatic and non-toothed clamps used to clamp blood vessels, grab tissue or vessels. They are often known as a "snap."
Catalog No. 50-253-8285 Supplier World Precision Instrument Supplier No. WP509970
Crile hemostatic forceps are atraumatic and non-toothed clamps used to clamp blood vessels, grab tissue or vessels. They are often known as a "snap."