Learn More
World Precision Instrument Crile Hemostatic Forceps, 14 cm, Straight, Smooth
SDP
Catalog No. 502521284
Encompass
Crile hemostatic forceps are atraumatic and non-toothed clamps used to clamp blood vessels, grab tissue or vessels. They are often known as a "snap."
Catalog No. 50-252-1284 Supplier World Precision Instrument Supplier No. 503227
Crile hemostatic forceps are atraumatic and non-toothed clamps used to clamp blood vessels, grab tissue or vessels. They are often known as a "snap."