Learn More
Sklar Instruments Crile-Wood Needle Holder, Sklar, 7 in., Blunt, Straight, Cross-Serrated with Groove, Stainless Steel. EA/1.
SDP
Catalog No. 501437447 Shop All Sklar Instruments Products
Encompass_Preferred
Crile-Wood Needle Holder, Sklar, 7 in., Blunt, Straight, Cross-Serrated with Groove, Stainless Steel. Sklar's Crile-Wood Needle Holder is used to hold and guide small to medium size needles and suture materials. The Crile-Wood Needle Holder has a gently tapered, blunt tip and is generally more delicate than the Mayo-Hegar but slightly less delicate than Webster, Halsey, or DeBakey needle holders. The ratcheted finger ring handle aids in securing needles for both continuous and interrupted closures. This product is straight with cross-serrated tips, center notch and an overall length of 7 inches.
This item is not returnable. View return policy
Catalog No. 50-143-7447 Supplier Sklar Instruments Supplier No. 201270
Crile-Wood Needle Holder, Sklar, 7 in., Blunt, Straight, Cross-Serrated with Groove, Stainless Steel. Sklar's Crile-Wood Needle Holder is used to hold and guide small to medium size needles and suture materials. The Crile-Wood Needle Holder has a gently tapered, blunt tip and is generally more delicate than the Mayo-Hegar but slightly less delicate than Webster, Halsey, or DeBakey needle holders. The ratcheted finger ring handle aids in securing needles for both continuous and interrupted closures. This product is straight with cross-serrated tips, center notch and an overall length of 7 inches.