Learn More
Fine Science Tools CRILE-WOOD NEEDLE HOLDER 15CM
SDP
Catalog No. NC1563401
Encompass
Product: Crile-Wood Needle Holder/15cm/ Tip Shape: Straight/ Tip Width: 1.5mm/ Clamping Length: 13mm/ Scissors: No/ Lock: Yes/ Alloy: Stainless Steel/ Length: 15cm/ Serrated: Yes/ Unit: Each
Catalog No. NC1563401 Supplier Fine Science Tools Supplier No. 1200315
Product: Crile-Wood Needle Holder/15cm/ Tip Shape: Straight/ Tip Width: 1.5mm/ Clamping Length: 13mm/ Scissors: No/ Lock: Yes/ Alloy: Stainless Steel/ Length: 15cm/ Serrated: Yes/ Unit: Each