Learn More
Med Vet International Crile-Wood Needle Holder, 6-in. (15.24cm) Tungsten Carbide, J0078X
Catalog No. 5000584167
Encompass_Preferred
Finer, narrower jaws than the Mayo-Hegar.
Catalog No. 50-005-84167 Supplier Med Vet International Supplier No. JOR78X
Finer, narrower jaws than the Mayo-Hegar.