Learn More
Med Vet International Miltex Vantage Crile-Wood Needle Holder, 6in., Delicate Jaws
Catalog No. 502831799
Encompass_Preferred
The Miltex Vantage V98-50 Crile-Wood needle holder is made from high quality Japanese stainless steel and measures 6" in length. It has a finger ring handle with a ratchet lock and straight, serrated jaws.
Catalog No. 50-283-1799 Supplier Med Vet International Supplier No. MILV9850
The Miltex Vantage V98-50 Crile-Wood needle holder is made from high quality Japanese stainless steel and measures 6" in length. It has a finger ring handle with a ratchet lock and straight, serrated jaws.