Learn More
Sklar Instruments Surgi-OR TC Crile-Wood Needle Holder, Sklar 8 in., Straight, Cross-Serrated, Tungsten Carbide / Stainless Steel. EA.
SDP
Catalog No. 501184173 Shop All Sklar Instruments Products
Encompass_Preferred
Surgi-OR TC Crile-Wood Needle Holder, Sklar. 8 in., Straight, Cross-Serrated, Tungsten Carbide / Stainless Steel. The Surgi-OR TC Crile-Wood Needle Holder is used to hold and guide small to medium size needles and suture materials. The Crile-Wood Needle Holder has a gently tapered, blunt tip and is generally more delicate than the Mayo-Hegar but slightly less delicate than Webster, Halsey, or DeBakey needle holders. The ratcheted finger ring handle aids in securing needles for both continuous and interrupted closures. Hand-crafted in Pakistan, Surgi-OR TC mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. They are designed to stay sharp for a significantly longer period of time than other standard instruments. This product is straight with cross-serrated tips, and an overall length of 8 inches.
Catalog No. 50-118-4173 Supplier Sklar Instruments Supplier No. 95139
Surgi-OR TC Crile-Wood Needle Holder, Sklar. 8 in., Straight, Cross-Serrated, Tungsten Carbide / Stainless Steel. The Surgi-OR TC Crile-Wood Needle Holder is used to hold and guide small to medium size needles and suture materials. The Crile-Wood Needle Holder has a gently tapered, blunt tip and is generally more delicate than the Mayo-Hegar but slightly less delicate than Webster, Halsey, or DeBakey needle holders. The ratcheted finger ring handle aids in securing needles for both continuous and interrupted closures. Hand-crafted in Pakistan, Surgi-OR TC mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. They are designed to stay sharp for a significantly longer period of time than other standard instruments. This product is straight with cross-serrated tips, and an overall length of 8 inches.