Learn More
AMPLYUS LLC miniPCR bio Cystic Fibrosis Lab: Missy Baker Missing™
SDP
Catalog No. NC1243151
Encompass_Preferred
Missy Baker is missing and police have two suspects. Knowing that Missy Baker has cystic fibrosis, students test DNA evidence from both suspects' cars to see if it matches a person with a CFTR gene deletion, introducing PCR and gel electrophoresis techniques.
Catalog No. NC1243151 Supplier AMPLYUS LLC Supplier No. KT100003
Missy Baker is missing and police have two suspects. Knowing that Missy Baker has cystic fibrosis, students test DNA evidence from both suspects' cars to see if it matches a person with a CFTR gene deletion, introducing PCR and gel electrophoresis techniques.