Learn More
Description
- Available with numbering from 1 to 8 or 9 to 16.
- Bright orange in color
- Withstands high winds
Specifications
Specifications
| Product Type | Crime Scene Cone |
| Includes | Numbers 1 to 8 |
| For Use With (Application) | Used at Crime Scenes |
| Color | Orange, Black |
| Quantity | 8/Cs. |
| Length (English) | 6 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
