Learn More
HORIBA INSTRUMENTS INCORPORATED CrimeScope CS-16-500W Forensic Light Source.Laboratory Unit – 15 tiltable wavelengths + IR port.
Catalog No. NC3136499
Encompass
500W Xenon lamp with 1000+ hour lifetime.The CrimeScope is offered with 16 wavelengths: UV 300-400nm, 415nm, 445nm, 455nm, 475nm, 495nm, CSS (Crime Scene Search shortpass filter SP540nm), 515nm, 535nm, 555nm, SP575nm, 600nm, 630nm, 670nm, white light and infrared output.Delivered with complete set of accessories:- 10mm dia. dual armored liquid light guide,- Push-button remote control integrated at working end of light guide,- 4 pairs of goggles (UV, YW, OR, RD),- 3-62mm Camera Filters (YW, Orange, Red)- 1 flexible arm and 2 rugged clamps,- Manual
Catalog No. NC3136499 Supplier HORIBA INSTRUMENTS INCORPORATED Supplier No. 5500000388
Add to Cart
Add to Cart
Request a Quote
500W Xenon lamp with 1000+ hour lifetime.The CrimeScope is offered with 16 wavelengths: UV 300-400nm, 415nm, 445nm, 455nm, 475nm, 495nm, CSS (Crime Scene Search shortpass filter SP540nm), 515nm, 535nm, 555nm, SP575nm, 600nm, 630nm, 670nm, white light and infrared output.Delivered with complete set of accessories:- 10mm dia. dual armored liquid light guide,- Push-button remote control integrated at working end of light guide,- 4 pairs of goggles (UV, YW, OR, RD),- 3-62mm Camera Filters (YW, Orange, Red)- 1 flexible arm and 2 rugged clamps,- Manual