Learn More
Description
- Features designated areas for pertinent information and is printed on both the front and back in red.
- Made from durable 110 lb. index cardstock.
- Compatible with any type of fingerprinting ink.
- Flexible and durable construction.
- Resists creasing and wrinkling.
Specifications
Specifications
| Dimensions (L x W) | 8 x 8 in. |
| Product Type | Fingerprint Record Card |
| For Use With (Application) | Fingerprinting Criminals |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
