Learn More
Krackeler Scientific Crimp Caps
SDP
Catalog No. 056005241N
Change view
Click to view available options
Material (Liner):
PTFE
PTFE/Butyl
PTFE/Red Rubber
PTFE/Silicone
Closure Size:
11 mm
20 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material (Liner) Closure Size
056005241N PTFE/Silicone 20 mm
HP51834479N PTFE/Butyl 20 mm
51820552N PTFE 11 mm
HP51811210N PTFE/Red Rubber 11 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 056005241N Supplier Krackeler Scientific Supplier No. 9251834477EA
Add to Cart
Add to Cart
For headspace top vials
Specifications
Material (Liner) PTFE/Silicone
Color Silver
Material Aluminum
Quantity 100 Pk.
Closure Size 20 mm
Product Type Crimp Cap