Learn More
Krackeler Scientific Crimp Caps
Description
- Supplied as a cap and septum assembly.
- Intended for chromatographic sample containment.
Specifications
Specifications
| Material (Liner) | PTFE/Butyl |
| Color | Silver |
| Material | Aluminum |
| Pre-slit | No |
| Septum | PTFE/Butyl |
| Quantity | 100 Pk. |
| Closure Size | 20 mm |
| Product Type | Crimp Cap |